Learn More
Cayman Chemical L-Sulforaphn 10mg
SDP
Catalog No. 502968899
Encompass_Preferred
An isothiocyanate derived from glucoraphanin; powerful antioxidant, anti-inflammatory, and anti-carcinogenic effects; at 40 µM, activates Nrf2-mediated gene expression of phase II detoxification enzymes by disrupting the Keap1-Nrf2 complex, allowing Nrf2 to alter transcription
Catalog No. 50-296-8899 Supplier Cayman Chemical Supplier No. 1479710 MG
An isothiocyanate derived from glucoraphanin; powerful antioxidant, anti-inflammatory, and anti-carcinogenic effects; at 40 µM, activates Nrf2-mediated gene expression of phase II detoxification enzymes by disrupting the Keap1-Nrf2 complex, allowing Nrf2 to alter transcription