Learn More
Revvity Health Sciences Inc L-[35S]-Methionine, 10mCi (370MBq), Specific Activity:>1000Ci (37.0TBq)/mmole, 50mM Tricine, 10mM BME
Catalog No. 502809552
Encompass
L-[35S]-Methionine, 10mCi (370MBq), Specific Activity:>1000Ci (37.0TBq)/mmole, 50mM Tricine, 10mM BME
Catalog No. 50-280-9552 Supplier Revvity Health Sciences Inc Supplier No. NEG009C010MC
Add to Cart
Add to Cart
Request a Quote
L-[35S]-Methionine, 10mCi (370MBq), Specific Activity:>1000Ci (37.0TBq)/mmole, 50mM Tricine, 10mM BME