Learn More
Revvity Health Sciences Inc L-[35S]-Methionine, 1mCi (37MBq), Specific Activity:>800Ci (29.6TBq)/mmole, 50mM Tricine, 10mM BME
Catalog No. 502810790
Encompass
L-[35S]-Methionine, 1mCi (37MBq), Specific Activity:>800Ci (29.6TBq)/mmole, 50mM Tricine, 10mM BME
Catalog No. 50-281-0790 Supplier Revvity Health Sciences Inc Supplier No. NEG009H001MC
L-[35S]-Methionine, 1mCi (37MBq), Specific Activity:>800Ci (29.6TBq)/mmole, 50mM Tricine, 10mM BME