Learn More
Andwin Scientific L.T.II COVERERGLASS 16/PK
SDP
Catalog No. 50930229
Encompass_Preferred
Specimen Preparation & Embedding Supplies: Lab-Tek Chamber Slide Products: Lab-Tek Chambered Coverglass System: Lab-Tek II Chamber Coverglass: L.T. II Coverglass, 8 Chambers, 16/pk
Catalog No. 50-930-229 Supplier Andwin Scientific Supplier No. 7037781
Specimen Preparation & Embedding Supplies: Lab-Tek Chamber Slide Products: Lab-Tek Chambered Coverglass System: Lab-Tek II Chamber Coverglass: L.T. II Coverglass, 8 Chambers, 16/pk