Learn More
Alkali Scientific L (+) Tartaric Acid [(2R, 3R) (+) Tartaric acid/ L Threaric acid], 500 G
SDP
Catalog No. 5000661102
Encompass_Preferred
L (+) Tartaric Acid, ACS Grade, is a high-purity organic acid and chiral compound, available in a 500g quantity. It is widely used in laboratory settings for various chemical reactions and is particularly important in the synthesis of enantiomerically pure compounds. Tartaric acid serves as a reagent in asymmetric synthesis and as a key intermediate in the production of other chiral molecules. It is also commonly used in optical isomer studies, making it essential for applications in pharmacology, biochemistry, and materials science. This laboratory-grade tartaric acid is highly pure and ideal for both academic and industrial research applications.
Catalog No. 50-006-61102 Supplier Alkali Scientific Supplier No. LTA001CP
L (+) Tartaric Acid, ACS Grade, is a high-purity organic acid and chiral compound, available in a 500g quantity. It is widely used in laboratory settings for various chemical reactions and is particularly important in the synthesis of enantiomerically pure compounds. Tartaric acid serves as a reagent in asymmetric synthesis and as a key intermediate in the production of other chiral molecules. It is also commonly used in optical isomer studies, making it essential for applications in pharmacology, biochemistry, and materials science. This laboratory-grade tartaric acid is highly pure and ideal for both academic and industrial research applications.