Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich L-(+)-Tartaric Acid

Catalog No. 503081424 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1424 500 g
25-138-025KG 2.5 kg
25-138-05G 5 g
25-138-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-308-1424 Supplier Sigma Aldrich Supplier No. 251380500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

L-(+)-Tartaric Acid

Specifications

CAS 87-69-4
Melting Point 170°C to 172°C
Molecular Formula C4H6O6
Linear Formula HO2CCH(OH)CH(OH)CO2H
MDL Number MFCD00064207
Quantity 500 g
Synonym (2R,3R)-(+)-Tartaric acid; L-Threaric acid
Molecular Weight (g/mol) 150.09 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.