Learn More
Selleck Chemical LLC L-Tartaric acid
SDP
Catalog No. 5000002940
Encompass_Preferred
L-Tartaric acid (2 3-Dihydroxysuccinic acid Threaric acid Racemic acid Uvic acid Paratartaric acid Winestone) is a white crystalline organic acid that occurs naturally in many plants most notably in grapes It has several applications for industrial and pharmaceutical use
Catalog No. 50-000-02940 Supplier Selleck Chemical LLC Supplier No. S623325MG
L-Tartaric acid (2 3-Dihydroxysuccinic acid Threaric acid Racemic acid Uvic acid Paratartaric acid Winestone) is a white crystalline organic acid that occurs naturally in many plants most notably in grapes It has several applications for industrial and pharmaceutical use