Learn More
Bioworld L-(+)-Tartaric Acid, 10% (w/v), 1 L
Catalog No. 50255637
Encompass_Preferred
L-(+)-Tartaric Acid Solution 10% (w/v) in biotechnology grade water.Appearance: LiquidBoiling Point : 399.3°CDensity : ~1.9 g/cm3 (Predicted)Melting Point: 170-172 °C(lit.)RTECS Number: WW7875000
Catalog No. 50-255-637 Supplier Bioworld Supplier No. 402201502
L-(+)-Tartaric Acid Solution 10% (w/v) in biotechnology grade water.Appearance: LiquidBoiling Point : 399.3°CDensity : ~1.9 g/cm3 (Predicted)Melting Point: 170-172 °C(lit.)RTECS Number: WW7875000