Learn More
Cayman Chemical L-Tetrahydropalmatine, Purity: ≥98%, CAS: 483-14-7, 25 mg
SDP
Catalog No. 502983045
Encompass_Preferred
An alkaloid that antagonizes the supraspinal dopamine 2 receptor and reduces stress-induced anxiety in rats
Catalog No. 50-298-3045 Supplier Cayman Chemical Supplier No. 2053525 MG
An alkaloid that antagonizes the supraspinal dopamine 2 receptor and reduces stress-induced anxiety in rats