Learn More
Enzo Life Sciences L-Theanine (25g). CAS: 3081-61-6
SDP
Catalog No. 502007747
Encompass_Preferred
Amino acid analog of glutamine, uniquely found in green tea. Increases brain serotonin, dopamine and GABA levels and binds to AMPA, kainate and NMDA receptors. Has been shown to exert neuroprotective effects in animal models possibly through its antagonistic effects on group 1 metabotrophic glutamate receptors. Behavioural studies with rats suggest L-Theanine to positiveley affect learning and memory processes. Promotes self-renewal of human embryonic stem cells. Alternative name: Ngamma-Ethyl-L-glutamine, L-Glutamic acid gamma-(ethylamide). Formula: C7H14N2O3. MW: 174.20. Purity: ≥98% (Titration). Appearance: White to off-white solid. Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C. Handling: Protect from moisture.
Catalog No. 50-200-7747 Supplier Enzo Life Sciences Supplier No. ALX106042G025
Amino acid analog of glutamine, uniquely found in green tea. Increases brain serotonin, dopamine and GABA levels and binds to AMPA, kainate and NMDA receptors. Has been shown to exert neuroprotective effects in animal models possibly through its antagonistic effects on group 1 metabotrophic glutamate receptors. Behavioural studies with rats suggest L-Theanine to positiveley affect learning and memory processes. Promotes self-renewal of human embryonic stem cells. Alternative name: Ngamma-Ethyl-L-glutamine, L-Glutamic acid gamma-(ethylamide). Formula: C7H14N2O3. MW: 174.20. Purity: ≥98% (Titration). Appearance: White to off-white solid. Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C. Handling: Protect from moisture.