Learn More
Lkt Laboratories L-Theanine, ≥98%, 1GR
SDP
Supplier:  Lkt Laboratories T28161G
Encompass_Preferred
Non-essential amino acid found in Camellia and Boletus, glutamic acid analog; weak AMPA and NMDA agonist. CAS: 3081-61-6. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4934