Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Theanine, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
3081-61-6
C7H14N2O3
174.2
DATAGRPVKZEWHA-YFKPBYRVSA-N
l-theanine,theanine,theanin,s-2-amino-5-ethylamino-5-oxopentanoic acid,n-ethyl-l-glutamine,suntheanine,n5-ethyl-l-glutamine,n 5-ethyl-l-glutamine,unii-8021pr16qo
439378
CHEBI:17394
(2S)-2-amino-5-(ethylamino)-5-oxopentanoic acid
CCNC(=O)CCC(C(=O)O)N
3081-61-6
C7H14N2O3
174.2
DATAGRPVKZEWHA-YFKPBYRVSA-N
l-theanine,theanine,theanin,s-2-amino-5-ethylamino-5-oxopentanoic acid,n-ethyl-l-glutamine,suntheanine,n5-ethyl-l-glutamine,n 5-ethyl-l-glutamine,unii-8021pr16qo
439378
CHEBI:17394
(2S)-2-amino-5-(ethylamino)-5-oxopentanoic acid
CCNC(=O)CCC(C(=O)O)N
Melting Point 206.0°C to 219.0°C
Infrared Spectrum Authentic
Quantity 5g
Linear Formula CH3CH2NHC(=O)CH2CH2CH(NH2)C(=O)OH
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in ether, alcohols
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material L-Theanine

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 221-379-