Learn More
Medchemexpress LLC L-theanine (standard) | 3081-61-6 | C7H14N2O3 | 50 MG
SDP
Catalog No. 5000375809
Encompass_Preferred
L-Theanine (Standard) is the analytical standard of L-Theanine, a non-protein amino acid found in green tea leaves. Intended for research and analytical applications, it blocks the binding of L-glutamic acid to glutamate receptors in the brain. This compound possesses neuroprotective, anticancer, and anti-oxidative activities, and is orally active with the ability to cross the blood-brain barrier.
  • Used as an analytical standard and reference for assays
  • Ideal for qualitative, quantitative, and methodological research (e.g., HPLC, GC, MS)
  • Exhibits neuroprotective, anticancer, and anti-oxidative activities
  • Blocks L-glutamic acid binding to glutamate receptors
  • Orally active and can cross the blood-brain barrier
Catalog No. 50-003-75809 Supplier Medchemexpress LLC Supplier No. HY15121R50MG
L-Theanine (Standard) is the analytical standard of L-Theanine, a non-protein amino acid found in green tea leaves. Intended for research and analytical applications, it blocks the binding of L-glutamic acid to glutamate receptors in the brain. This compound possesses neuroprotective, anticancer, and anti-oxidative activities, and is orally active with the ability to cross the blood-brain barrier.
  • Used as an analytical standard and reference for assays
  • Ideal for qualitative, quantitative, and methodological research (e.g., HPLC, GC, MS)
  • Exhibits neuroprotective, anticancer, and anti-oxidative activities
  • Blocks L-glutamic acid binding to glutamate receptors
  • Orally active and can cross the blood-brain barrier