Learn More
Medchemexpress LLC L-threo-3-hydroxyaspartic acid | 7298-99-9 | MFCD00673765 | 98.8% | 149.10 g/mol | C4H7NO5 | 10 MG
SDP
Catalog No. 5000211941
Encompass_Preferred
L-threo-3-Hydroxyaspartic acid is a research-grade amino acid derivative that functions as a potent inhibitor of excitatory amino acid transporters (EAATs). Supplied as a powder for biochemical and cellular studies, it has been characterized in HEK293 cell lines and includes batch-specific purity and handling information in accompanying certificates of analysis and SDS.
Catalog No. 50-002-11941 Supplier Medchemexpress LLC Supplier No. HY10080110MG
L-threo-3-Hydroxyaspartic acid is a research-grade amino acid derivative that functions as a potent inhibitor of excitatory amino acid transporters (EAATs). Supplied as a powder for biochemical and cellular studies, it has been characterized in HEK293 cell lines and includes batch-specific purity and handling information in accompanying certificates of analysis and SDS.