Learn More
Medchemexpress LLC L-threo-3-hydroxyaspartic acid | 7298-99-9 | MFCD00673765 | 98.8% | 149.10 g/mol | C4H7NO5 | 5 MG

Supplier: Medchemexpress LLC HY1008015MG
L-threo-3-Hydroxyaspartic acid is a research-grade small molecule that inhibits excitatory amino acid transporters (EAATs). Supplied as a solid powder for biochemical and cellular studies, it exhibits micromolar inhibitory activity in HEK293 cell assays.
- Potent EAAT inhibitor with Ki values of 11 μM (EAAT1), 19 μM (EAAT2), and 14 μM (EAAT3).
- High purity: 98.8% suitable for research applications.
- Powder form stable for long-term storage at -20°C; in solution store at -80°C for extended periods.
- Molecular formula C4H7NO5 and molecular weight 149.10 g/mol.
- Useful tool compound for studies of glutamate transport and synaptic physiology.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.