Learn More
Abcam L-threo-PDMP, 10MG
Catalog No. 502706043
Encompass_Preferred
MW 427.03 g/mol, Purity >98%. Enantiomer of D-threo-PDMP (Asc 4052). Activates glycosyltransferases for ganglioside biosynthesis. Antineurodegenerative properties. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6043 Supplier Abcam Supplier No. AB14404610MG
MW 427.03 g/mol, Purity >98%. Enantiomer of D-threo-PDMP (Asc 4052). Activates glycosyltransferases for ganglioside biosynthesis. Antineurodegenerative properties. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement