Learn More
Cayman Chemical L-threo-PPMPhydrochlorIde 10mg
SDP
Catalog No. 502934418
Encompass_Preferred
An inhibitor of glucosylceramide synthetase; isomer of D-threo-PPMP; inhibits the growth of MCF-7, MDA-MB-468, and SK-BR-3 breast cancer cells (ED50s = 4, 7, and 6 μM, respectively)
Catalog No. 50-293-4418 Supplier Cayman Chemical Supplier No. 2452510 MG
An inhibitor of glucosylceramide synthetase; isomer of D-threo-PPMP; inhibits the growth of MCF-7, MDA-MB-468, and SK-BR-3 breast cancer cells (ED50s = 4, 7, and 6 μM, respectively)