Learn More
Medchemexpress LLC L-threonic acid magnesium | 778571-57-6 | MFCD16621117 | ≥99.0% | 294.50 g/mol | C8H14MgO10 | 5 MG
SDP
Catalog No. 5000223340
Encompass_Preferred
L-threonic acid magnesium (magnesium L-threonate) is the magnesium salt of L-threonic acid, used in research to increase brain magnesium levels and to study neuroinflammation and cognitive disorders. It is orally active and supplied as a high-purity solid suitable for in vitro and in vivo studies.
  • Magnesium salt of L-threonic acid, chemical formula C8H14MgO10.
  • Orally active and used in models of Alzheimer's disease and neuroinflammation.
  • Reported to increase brain magnesium concentration.
  • High purity (≥99.0%) suitable for research applications.
  • Stable when stored sealed at 4°C; in solution: -80°C (6 months), -20°C (1 month).
  • SDS available in multiple languages.
Catalog No. 50-002-23340 Supplier Medchemexpress LLC Supplier No. HY1342405MG
L-threonic acid magnesium (magnesium L-threonate) is the magnesium salt of L-threonic acid, used in research to increase brain magnesium levels and to study neuroinflammation and cognitive disorders. It is orally active and supplied as a high-purity solid suitable for in vitro and in vivo studies.
  • Magnesium salt of L-threonic acid, chemical formula C8H14MgO10.
  • Orally active and used in models of Alzheimer's disease and neuroinflammation.
  • Reported to increase brain magnesium concentration.
  • High purity (≥99.0%) suitable for research applications.
  • Stable when stored sealed at 4°C; in solution: -80°C (6 months), -20°C (1 month).
  • SDS available in multiple languages.