Learn More
Medchemexpress LLC L-threonic acid (magnesium) | 778571-57-6 | MFCD16621117 | ≥99.0% | C8H14MgO10 | 10MG
SDP
Catalog No. 5000259033
Encompass_Preferred
L-Threonic acid magnesium (magnesium L-threonate) is the magnesium salt of L-threonic acid supplied as a white to off-white solid for research use. It is used in preclinical studies to modulate magnesium levels and investigate neuroinflammation and cognitive effects. Formula: C8H14MgO10; molecular weight: 294.50; provided at high purity for experimental applications.
  • High purity (≥99.0%).
  • Water solubility achievable with ultrasonic warming (4.17 mg/mL).
  • White to off-white solid appearance.
  • Recommended storage at 4°C, sealed and protected from moisture.
  • Suitable for preclinical research into magnesium bioavailability and neuroinflammation.
Catalog No. 50-002-59033 Supplier Medchemexpress LLC Supplier No. HY13424010MG
L-Threonic acid magnesium (magnesium L-threonate) is the magnesium salt of L-threonic acid supplied as a white to off-white solid for research use. It is used in preclinical studies to modulate magnesium levels and investigate neuroinflammation and cognitive effects. Formula: C8H14MgO10; molecular weight: 294.50; provided at high purity for experimental applications.
  • High purity (≥99.0%).
  • Water solubility achievable with ultrasonic warming (4.17 mg/mL).
  • White to off-white solid appearance.
  • Recommended storage at 4°C, sealed and protected from moisture.
  • Suitable for preclinical research into magnesium bioavailability and neuroinflammation.