Learn More
TARGETMOL CHEMICALS INC L-Threonic acid magnesium salt
SDP
Catalog No. 502877210
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. L-Threonic acid magnesium salt (L-TAMS) increases synapse density and memory ability in both aged rats and late stage Alzheimer’s disease (AD) model mice. Purity 99.73%
Catalog No. 50-287-7210 Supplier TARGETMOL CHEMICALS INC Supplier No. T937950MG
Also available in 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. L-Threonic acid magnesium salt (L-TAMS) increases synapse density and memory ability in both aged rats and late stage Alzheimer’s disease (AD) model mice. Purity 99.73%