Learn More
Cayman Chemical L-ThreonIn-d2 10mg
SDP
Catalog No. 502910468
Encompass_Preferred
An internal standard for the quantification of L-threonine by GC- or LC-MS
Catalog No. 50-291-0468 Supplier Cayman Chemical Supplier No. 3482810 MG
An internal standard for the quantification of L-threonine by GC- or LC-MS