Learn More
Cayman Chemical L-ThreonIn-d2 1mg
SDP
Catalog No. 502911075
Encompass_Preferred
An internal standard for the quantification of L-threonine by GC- or LC-MS
Catalog No. 50-291-1075 Supplier Cayman Chemical Supplier No. 348281 MG
An internal standard for the quantification of L-threonine by GC- or LC-MS