Learn More
Cayman Chemical L-Threonine-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 1202936-45-5, 25 mg
SDP
Catalog No. 502912733
Encompass_Preferred
An internal standard for the quantification of L-threonine by GC- or LC-MS
Catalog No. 50-291-2733 Supplier Cayman Chemical Supplier No. 3482825 MG
An internal standard for the quantification of L-threonine by GC- or LC-MS