Learn More
Cayman Chemical L-Threonine-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 1202936-45-5, 5 mg
SDP
Catalog No. 502911308
Encompass_Preferred
An internal standard for the quantification of L-threonine by GC- or LC-MS
Catalog No. 50-291-1308 Supplier Cayman Chemical Supplier No. 348285 MG
An internal standard for the quantification of L-threonine by GC- or LC-MS