Learn More
Medchemexpress LLC L-Threonine-13C4 | 55443-53-3 | 123.09 | 5 MG
SDP
Catalog No. 5000388483
Encompass_Preferred
L-Threonine-13C4 is the 13C labeled L-Threonine. L-Threonine is a natural amino acid, produced by microbial fermentation, and used in food, medicine, or feed.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
  • Has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88483 Supplier Medchemexpress LLC Supplier No. HYN0658S65MG
L-Threonine-13C4 is the 13C labeled L-Threonine. L-Threonine is a natural amino acid, produced by microbial fermentation, and used in food, medicine, or feed.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
  • Has the potential to affect the pharmacokinetic and metabolic profiles of drugs.