Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Threonine-d2 | 1202936-45-5 | 121.13 | 1 MG
SDP

Catalog No. 5000388541
Encompass_Preferred

L-Threonine-d2 is the deuterium-labeled form of L-Threonine, a natural amino acid. It is produced through microbial fermentation and finds applications in food, medicine, or animal feed. This compound is intended for research use only and is not sold to patients.

  • Deuterium labeled L-Threonine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.76%.
  • Appearance: Solid, white to off-white.

Catalog No. 50-003-88541 Supplier Medchemexpress LLC Supplier No. HYN0658S51MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Threonine-d2 is the deuterium-labeled form of L-Threonine, a natural amino acid. It is produced through microbial fermentation and finds applications in food, medicine, or animal feed. This compound is intended for research use only and is not sold to patients.

  • Deuterium labeled L-Threonine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.76%.
  • Appearance: Solid, white to off-white.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.