Learn More
Medchemexpress LLC L-Threonine-d5 | 1217456-38-6 | CB71121697 | 124.15 | 5 MG
SDP
Catalog No. 5000388660
Encompass_Preferred
L-Threonine-d5 is the deuterium labeled L-Threonine, a natural amino acid produced by microbial fermentation and used in food, medicine, or feed applications. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers for quantitation during drug development, potentially influencing pharmacokinetic and metabolic profiles. This compound can serve as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium labeling can influence drug pharmacokinetics and metabolic profiles
  • Produced by microbial fermentation
Catalog No. 50-003-88660 Supplier Medchemexpress LLC Supplier No. HYN0658S15MG
L-Threonine-d5 is the deuterium labeled L-Threonine, a natural amino acid produced by microbial fermentation and used in food, medicine, or feed applications. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers for quantitation during drug development, potentially influencing pharmacokinetic and metabolic profiles. This compound can serve as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium labeling can influence drug pharmacokinetics and metabolic profiles
  • Produced by microbial fermentation