Learn More
Medchemexpress LLC L-threonine, L-phenylalanyl- | 51352-44-4 | 95.8% | 266.29 g/mol | C13H18N2O4 | 5mg
SDP
Catalog No. 5000269594
Encompass_Preferred
L-Phenylalanyl-L-threonine is a dipeptide containing Phenylalanine and threonine L-Phenylalanyl-L-threonine can be used to synthesize the cyclic hexapeptide Banyascyclamide A[1]
Catalog No. 50-002-69594 Supplier Medchemexpress LLC Supplier No. HYW9672235MG
L-Phenylalanyl-L-threonine is a dipeptide containing Phenylalanine and threonine L-Phenylalanyl-L-threonine can be used to synthesize the cyclic hexapeptide Banyascyclamide A[1]