Learn More
Medchemexpress LLC L-threonine, N-[[[1S-3-amino-1-[3-[1R-1-amino-2-hydroxyethyl]-1,2,4-oxadiazol-5-yl]-3-oxopropyl] | 1673534-76-3 | 360.32 | C12H20N6O7 | 100 MG
SDP
Catalog No. 5000211835
Encompass_Preferred
CA-170 is a small-molecule dual inhibitor of the immune checkpoints VISTA and PD-L1 developed for preclinical research. It modulates T-cell proliferation and effector functions suppressed by PD-L1/L2 and VISTA, and is used to study immune checkpoint pathways in vitro and in vivo.
  • Dual inhibitor of VISTA and PD-L1
  • Orally active small-molecule suitable for preclinical studies
  • Modulates T-cell proliferation and effector functions
  • Defined chemical properties to support reproducibility
  • Storage recommendations provided for powder and solutions
  • Intended for research use only
Catalog No. 50-002-11835 Supplier Medchemexpress LLC Supplier No. HY101093100MG
CA-170 is a small-molecule dual inhibitor of the immune checkpoints VISTA and PD-L1 developed for preclinical research. It modulates T-cell proliferation and effector functions suppressed by PD-L1/L2 and VISTA, and is used to study immune checkpoint pathways in vitro and in vivo.
  • Dual inhibitor of VISTA and PD-L1
  • Orally active small-molecule suitable for preclinical studies
  • Modulates T-cell proliferation and effector functions
  • Defined chemical properties to support reproducibility
  • Storage recommendations provided for powder and solutions
  • Intended for research use only