Learn More
Medchemexpress LLC L-threonine, N-[[[(1S)-3-amino-1-[3-[(1R)-1-amino-2-hydroxyethyl]-1,2,4-oxadiazol-5-yl]-3-oxopropyl)amino]carbonyl]- | 1673534-76-3 | 99.4% | 360.32 g·mol⁻¹ | C12H20N6O7 | 25 MG
SDP
Catalog No. 5000213515
Encompass_Preferred
CA-170 is a small-molecule, orally active dual inhibitor of the immune checkpoints VISTA and PD-L1, used as a research reagent to modulate immune responses in preclinical immunology and oncology studies. It has been shown to restore T-cell proliferation and effector functions inhibited by PD-L1 and VISTA and is supplied as a powder for laboratory use.
  • Oral small-molecule dual inhibitor of VISTA and PD-L1.
  • Restores T-cell proliferation and effector function in inhibited systems.
  • Suitable for in vitro and in vivo preclinical research applications.
  • High purity by HPLC (approximately 99.4%).
  • Molecular weight 360.32 g·mol⁻¹; formula C12H20N6O7.
  • Supplied as a powder with recommended cold-storage conditions.
Catalog No. 50-002-13515 Supplier Medchemexpress LLC Supplier No. HY10109325MG
CA-170 is a small-molecule, orally active dual inhibitor of the immune checkpoints VISTA and PD-L1, used as a research reagent to modulate immune responses in preclinical immunology and oncology studies. It has been shown to restore T-cell proliferation and effector functions inhibited by PD-L1 and VISTA and is supplied as a powder for laboratory use.
  • Oral small-molecule dual inhibitor of VISTA and PD-L1.
  • Restores T-cell proliferation and effector function in inhibited systems.
  • Suitable for in vitro and in vivo preclinical research applications.
  • High purity by HPLC (approximately 99.4%).
  • Molecular weight 360.32 g·mol⁻¹; formula C12H20N6O7.
  • Supplied as a powder with recommended cold-storage conditions.