Learn More
Selleck Chemical LLC L-Thyroxine (Levothyroxine) S2599-25mg
SDP
Catalog No. 503109825
Encompass_Preferred
L-Thyroxine (Levothyroxine) is a synthetic form of thyroxine and a hormone replacement drug L-Thyroxine can be used to induce animal models of Hyperthyroidism
Catalog No. 50-310-9825 Supplier Selleck Chemical LLC Supplier No. S259925MG
L-Thyroxine (Levothyroxine) is a synthetic form of thyroxine and a hormone replacement drug L-Thyroxine can be used to induce animal models of Hyperthyroidism