Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Thyroxine, 97+%, Thermo Scientific Chemicals

Catalog No. AC215662500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215662500 250 mg Glass bottle
AC215660010 1 g Glass bottle
AC215660050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC215662500 Supplier Thermo Scientific Chemicals Supplier No. 215662500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-48-9
Molecular Formula C15H11I4NO4
Molecular Weight (g/mol) 776.87
MDL Number MFCD00002595
InChI Key XUIIKFGFIJCVMT-UHFFFAOYNA-N
Synonym l-thyroxine, levothyroxine, thyroxine, thyroxin, synthroid, tetraiodothyronine, thyrax, levothyroxin, thyratabs, thyreoideum
PubChem CID 5819
ChEBI CHEBI:18332
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O

Specifications

Melting Point 235.0°C to 236.0°C
Color White to Beige
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Beilstein 14, IV, 2373
Merck Index 15, 9570
Specific Rotation Condition − 5.00 (20.00°C 1N NaOH:EtOH 1:2)
Specific Rotation − 5.00
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in alcohol and most other common, organic solvents, soluble in alcohol in the presence of mineral, acids or alkalies
Formula Weight 776.86
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material L-Thyroxine
Show More Show Less
EINECSNumber 200-101-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.