Learn More
Medchemexpress LLC L-Thyroxine (sodium salt pentahydrate) | 6106-07-6 | 99.9% | 888.93 | 1 ML
SDP
Catalog No. 5000387662
Encompass_Preferred
L-Thyroxine sodium salt pentahydrate (Levothyroxine; T4) is a synthetic hormone used in the research of hypothyroidism. DIO enzymes convert L-Thyroxine (T4) into the biologically active thyroid hormone Triiodothyronine (T3).
  • DIO enzymes activate thyroid hormone secretion
  • Modulates expression of ionic channels, pumps, and regulatory contractile proteins
  • Influences calcium homeostasis and flux
Catalog No. 50-003-87662 Supplier Medchemexpress LLC Supplier No. HY18341A10MM/1ML
L-Thyroxine sodium salt pentahydrate (Levothyroxine; T4) is a synthetic hormone used in the research of hypothyroidism. DIO enzymes convert L-Thyroxine (T4) into the biologically active thyroid hormone Triiodothyronine (T3).
  • DIO enzymes activate thyroid hormone secretion
  • Modulates expression of ionic channels, pumps, and regulatory contractile proteins
  • Influences calcium homeostasis and flux