Learn More
Medchemexpress LLC L-Thyroxine sodium xhydrate | 25416-65-3 | 98.9% | 816.87 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0106695G
Encompass_Preferred
L-Thyroxine sodium xhydrate (Levothyroxine; T4) is a synthetic hormone.
  • Used in the treatment of hypothyroidism.
  • DIO enzymes convert L-Thyroxine (T4) into biologically active hormone (Triiodothyronine, T3).
Catalog No. 50-004-51897