Learn More
Medchemexpress LLC L-tryptophan-1-13C | 81201-92-5 | MFCD06804672 | 99.0% | 205.22 g·mol⁻1 | C10(13C)H12N2O2 | 10 MG
SDP
Catalog No. 5000297060
Encompass_Preferred
L-Tryptophan-1-13C is the carbon-13 isotopically labeled form of the essential amino acid L-tryptophan, supplied as a white to off-white solid for use as an internal standard and tracer in metabolic, analytical, and spectroscopic studies. The product has a reported purity of 99.0% and is typically supplied in small-scale packs with recommended cold storage.
Catalog No. 50-002-97060 Supplier Medchemexpress LLC Supplier No. HYN0623S310MG
L-Tryptophan-1-13C is the carbon-13 isotopically labeled form of the essential amino acid L-tryptophan, supplied as a white to off-white solid for use as an internal standard and tracer in metabolic, analytical, and spectroscopic studies. The product has a reported purity of 99.0% and is typically supplied in small-scale packs with recommended cold storage.