Learn More
Medchemexpress LLC L-Tryptophan-α,β,β,2,4,5,6,7-d8 | 1233395-93-1 | 212.27 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S5100MG
Encompass_Preferred
L-Tryptophan-d8 is the deuterium-labeled L-Tryptophan. L-Tryptophan is an essential amino acid and a precursor of serotonin, melatonin, and vitamin B3.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88651