Learn More
Medchemexpress LLC L-Tryptophan-α,β,β,2,4,5,6,7-d8 | 1233395-93-1 | 212.27 | 25 MG
SDP
Catalog No. 5000390348
Encompass_Preferred
L-Tryptophan-d8 is the deuterium labeled L-Tryptophan. L-Tryptophan (Tryptophan) is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3. It is for research use only and not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules for quantitation during drug development
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-90348 Supplier Medchemexpress LLC Supplier No. 25MGHYN0623S5
L-Tryptophan-d8 is the deuterium labeled L-Tryptophan. L-Tryptophan (Tryptophan) is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3. It is for research use only and not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules for quantitation during drug development
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs