Learn More
Research Products International Corp L-Tryptophan, 500 Grams

Supplier: Research Products International Corp T60080500.0
CAS Number: 73-22-3
Molecular Formula: C11H12N2O2
Molecular Weight: 204.4
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.