Learn More
Medchemexpress LLC L-Tryptophan-d3 | 133519-78-5 | 207.24 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0623S91MG
Encompass_Preferred
L-Tryptophan-d3 is the deuterium labeled form of L-Tryptophan, an essential amino acid. L-Tryptophan acts as a precursor for vital compounds such as serotonin, melatonin, and vitamin B3. This compound serves as a tracer and an internal standard for quantitative analysis techniques like NMR, GC-MS, or LC-MS. Deuteration, which involves incorporating stable heavy isotopes, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Precursor for serotonin, melatonin, and vitamin B3.
  • Functions as a tracer in various studies.
  • Used as an internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, or LC-MS applications.
  • Deuteration may impact drug pharmacokinetic and metabolic profiles.
Catalog No. 50-003-88783