Learn More
Medchemexpress LLC L-Tryptophan-d3 | 133519-78-5 | 207.24 | 5 MG
SDP
Catalog No. 5000388518
Encompass_Preferred
L-Tryptophan-d3 is a deuterium labeled L-Tryptophan, an essential amino acid and a precursor of serotonin, melatonin, and vitamin B3. It is intended for research use only and not for sale to patients. This product can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-88518 Supplier Medchemexpress LLC Supplier No. HYN0623S95MG
L-Tryptophan-d3 is a deuterium labeled L-Tryptophan, an essential amino acid and a precursor of serotonin, melatonin, and vitamin B3. It is intended for research use only and not for sale to patients. This product can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS