Learn More
Medchemexpress LLC L-Tryptophan-d5 | 62595-11-3 | 209.26 | 100 MG
SDP
Catalog No. 5000388919
Encompass_Preferred
L-Tryptophan-d5 is a deuterium labeled L-Tryptophan, which is an essential amino acid and a precursor of serotonin, melatonin, and vitamin B3. Deuteration of drug molecules has garnered attention for its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can be incorporated into drug molecules for quantitation during drug development.
Catalog No. 50-003-88919 Supplier Medchemexpress LLC Supplier No. HYN0623S100MG
L-Tryptophan-d5 is a deuterium labeled L-Tryptophan, which is an essential amino acid and a precursor of serotonin, melatonin, and vitamin B3. Deuteration of drug molecules has garnered attention for its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can be incorporated into drug molecules for quantitation during drug development.