Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Tryptophan-d5 | 62595-11-3 | 209.26 | 25 MG
SDP

Catalog No. 5000389410
Encompass_Preferred

L-Tryptophan-d5 is a deuterium-labeled form of L-Tryptophan, an essential amino acid and a precursor to serotonin, melatonin, and vitamin B3. This product is intended for research use only.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Has a purity of 99.86% to 99.94%.
  • Contains deuterium, which can affect pharmacokinetic and metabolic profiles of drugs.
  • Appears as a solid with a yellow to orange color.
  • Store at 4°C, protected from light, under nitrogen.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light, under nitrogen.

Catalog No. 50-003-89410 Supplier Medchemexpress LLC Supplier No. 25MGHYN0623S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Tryptophan-d5 is a deuterium-labeled form of L-Tryptophan, an essential amino acid and a precursor to serotonin, melatonin, and vitamin B3. This product is intended for research use only.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Has a purity of 99.86% to 99.94%.
  • Contains deuterium, which can affect pharmacokinetic and metabolic profiles of drugs.
  • Appears as a solid with a yellow to orange color.
  • Store at 4°C, protected from light, under nitrogen.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light, under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.