Learn More
Medchemexpress LLC L-Tryptophan-d5 | 62595-11-3 | 209.26 | 50 MG
SDP
Catalog No. 5000388576
Encompass_Preferred
L-Tryptophan-d5 is a deuterium labeled form of L-Tryptophan, an essential amino acid and a precursor to serotonin, melatonin, and vitamin B3. This stable isotope can be utilized as a tracer in various studies or as an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. Deuteration may influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-88576 Supplier Medchemexpress LLC Supplier No. HYN0623S50MG
L-Tryptophan-d5 is a deuterium labeled form of L-Tryptophan, an essential amino acid and a precursor to serotonin, melatonin, and vitamin B3. This stable isotope can be utilized as a tracer in various studies or as an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. Deuteration may influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs