Learn More
Medchemexpress LLC L-Tryptophan-d5 | 62595-11-3 | MFCD28495036 | 209.26 | 1 MG
SDP
Catalog No. 5000388635
Encompass_Preferred
L-Tryptophan-d5 is a deuterium-labeled form of L-Tryptophan, an essential amino acid. It serves as a precursor for the synthesis of 5-HT, melatonin, and niacin, and is involved in various physiological processes.
  • Deuterium-labeled essential amino acid
  • Precursor for 5-HT, melatonin, and niacin synthesis
  • Involved in immune function
  • Plays a role in neuronal activity
Catalog No. 50-003-88635 Supplier Medchemexpress LLC Supplier No. HYN0623S1MG
L-Tryptophan-d5 is a deuterium-labeled form of L-Tryptophan, an essential amino acid. It serves as a precursor for the synthesis of 5-HT, melatonin, and niacin, and is involved in various physiological processes.
  • Deuterium-labeled essential amino acid
  • Precursor for 5-HT, melatonin, and niacin synthesis
  • Involved in immune function
  • Plays a role in neuronal activity