Learn More
Medchemexpress LLC L-Tryptophan-d8 | 1233395-93-1 | 212.27 | 1 MG
SDP
Catalog No. 5000388582
Encompass_Preferred
L-Tryptophan-d8 is the deuterium labeled L-Tryptophan. L-Tryptophan is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Useful for studying pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-88582 Supplier Medchemexpress LLC Supplier No. HYN0623S51MG
L-Tryptophan-d8 is the deuterium labeled L-Tryptophan. L-Tryptophan is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Useful for studying pharmacokinetic and metabolic profiles of drugs