Learn More
Medchemexpress LLC L-Tryptophan-d8 | 1233395-93-1 | 212.27 | 5 MG
SDP
Catalog No. 5000388583
Encompass_Preferred
L-Tryptophan-d8 is the deuterium labeled L-Tryptophan, an essential amino acid that acts as a precursor of serotonin, melatonin, and vitamin B3.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88583 Supplier Medchemexpress LLC Supplier No. HYN0623S55MG
L-Tryptophan-d8 is the deuterium labeled L-Tryptophan, an essential amino acid that acts as a precursor of serotonin, melatonin, and vitamin B3.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.