Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-tryptophan-d8 | 1233395-93-1 | 98.0% | 212.27 g/mol | C11H4D8N2O2 | 10 MG
SDP

Catalog No. 5000297994
Encompass_Preferred

L-Tryptophan-d8 is the deuterium-labeled form of the essential amino acid L-tryptophan, provided as a solid isotopic standard for analytical and metabolic applications. It is commonly used as an internal standard in mass spectrometry and tracer studies to distinguish labeled from endogenous tryptophan.

  • Deuterium labeled (d8).
  • Used as an isotopic internal standard for mass spectrometry and tracer studies.
  • Purity 98.0%.
  • Molecular formula C11H4D8N2O2; molecular weight 212.27 g/mol.
  • Physical form solid (powder), supplied as 10 mg unit.
  • Storage: 4°C for solid; in solution -80°C (up to 6 months) or -20°C (up to 1 month), sealed and protected from moisture and light.
  • CAS number 1233395-93-1.

Catalog No. 50-002-97994 Supplier Medchemexpress LLC Supplier No. HYN0623S510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Tryptophan-d8 is the deuterium-labeled form of the essential amino acid L-tryptophan, provided as a solid isotopic standard for analytical and metabolic applications. It is commonly used as an internal standard in mass spectrometry and tracer studies to distinguish labeled from endogenous tryptophan.

  • Deuterium labeled (d8).
  • Used as an isotopic internal standard for mass spectrometry and tracer studies.
  • Purity 98.0%.
  • Molecular formula C11H4D8N2O2; molecular weight 212.27 g/mol.
  • Physical form solid (powder), supplied as 10 mg unit.
  • Storage: 4°C for solid; in solution -80°C (up to 6 months) or -20°C (up to 1 month), sealed and protected from moisture and light.
  • CAS number 1233395-93-1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.