Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Tryptophan ethyl ester hydrochloride, 97%

Catalog No. AAH6695314
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6695314 25 g
AAH6695336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6695314 Supplier Thermo Scientific Chemicals Supplier No. H6695314

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

CAS: 2899-28-7 | C13H17ClN2O2 | 268.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2899-28-7
Molecular Formula C13H17ClN2O2
Molecular Weight (g/mol) 268.74
MDL Number MFCD00038993
InChI Key PESYCVVSLYSXAK-UHFFFAOYNA-N
Synonym l-tryptophan ethyl ester hydrochloride, ethyl l-tryptophanate hydrochloride, h-trp-oet.hcl, s-tryptophan ethyl ester hydrochloride, ethyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, h-trp-oet hcl, ethyl l-tryptophanate hcl, ethyl l-tryptophanate monohydrochloride, h-trp-oet. hcl, pubchem10920
PubChem CID 134519
IUPAC Name hydrogen ethyl 2-amino-3-(1H-indol-3-yl)propanoate chloride
SMILES [H+].[Cl-].CCOC(=O)C(N)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 220°C to 225°C
Color Brown
Beilstein 3919010
Quantity 25 g
Formula Weight 268.74
Percent Purity 97%
Physical Form Powder
Chemical Name or Material L-Tryptophan ethyl ester hydrochloride
EINECSNumber 220-786-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.