Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Tryptophan Ethyly Ester, >99%, MP Biomedicals™

Catalog No. ICN10315210 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10315210 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10315210 Supplier MP Biomedicals, Inc Supplier No. 0210315210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity research grade biochemical reagent for laboratory use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2899-28-7
Molecular Formula C13H17ClN2O2
Molecular Weight (g/mol) 268.74
MDL Number MFCD00038993
InChI Key PESYCVVSLYSXAK-UHFFFAOYNA-N
Synonym l-tryptophan ethyl ester hydrochloride, ethyl l-tryptophanate hydrochloride, h-trp-oet.hcl, s-tryptophan ethyl ester hydrochloride, ethyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, h-trp-oet hcl, ethyl l-tryptophanate hcl, ethyl l-tryptophanate monohydrochloride, h-trp-oet. hcl, pubchem10920
PubChem CID 134519
IUPAC Name hydrogen ethyl 2-amino-3-(1H-indol-3-yl)propanoate chloride
SMILES [H+].[Cl-].CCOC(=O)C(N)CC1=CNC2=CC=CC=C12

Specifications

Color White
Assay Percent Range > 99%
Quantity 10 g
Formula Weight 268.7
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material L-Tryptophan Ethyly Ester
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.