Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-tryptophan, N-[2-(1H-indol-3-yl)acetyl]- | 57105-53-0 | MFCD18641983 | 99.9% | 361.39 g·mol⁻1 | C21H19N3O3 | 5 MG
SDP

Catalog No. 5000223444
Encompass_Preferred

(S)-N-(1H-Indole-3-acetyl)tryptophan is a chiral tryptophan derivative supplied as a white to off-white solid for research use. It has been characterized by NMR and LCMS and is reported to weakly inhibit β-D-glucosidase, making it useful for biochemical and enzymology studies.

  • High purity: 99.9% (LCMS).
  • Well characterized by 1H NMR and LCMS.
  • Defined optical rotation: +11.00° (methanol, 25°C).
  • Molecular weight: 361.39 g·mol⁻1.
  • Recommended storage: powder at -20°C for long term stability.
  • Intended for research use only; not for human or diagnostic use.

Catalog No. 50-002-23444 Supplier Medchemexpress LLC Supplier No. HY1388155MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-N-(1H-Indole-3-acetyl)tryptophan is a chiral tryptophan derivative supplied as a white to off-white solid for research use. It has been characterized by NMR and LCMS and is reported to weakly inhibit β-D-glucosidase, making it useful for biochemical and enzymology studies.

  • High purity: 99.9% (LCMS).
  • Well characterized by 1H NMR and LCMS.
  • Defined optical rotation: +11.00° (methanol, 25°C).
  • Molecular weight: 361.39 g·mol⁻1.
  • Recommended storage: powder at -20°C for long term stability.
  • Intended for research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.